(4-Hydroxyphenyl)diphenyl methanol 98,0 HPLC_ASSAY_METHOD

Leverandør: TCI
Warning

Synonymer: 4-Hydroxytrityl Alcohol, 4-(Hydroxydiphenylmethyl)phenol

H1059-1G
TCIAH1059-1GEA 2625 NOK
TCIAH1059-1G
(4-Hydroxyphenyl)diphenyl methanol 98,0 HPLC_ASSAY_METHOD
(4-Hydroxyphenyl)diphenyl methanol
Formel: C₁₉H₁₆O₂
MW: 276.34 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00973626
CAS-nummer: 15658-11-4

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR