Biotin-LC-hydrazide ≥97.0% (by HPLC)

Leverandør: TCI

Synonymer: N''-Biotinyl-6-aminohexanoylhydrazide, N-(6-Hydrazinyl-6-oxohexyl)biotinamide, N-(6-Hydrazinyl-6-oxohexyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Biotinamidocaproyl hydrazine, Biotin LC hydrazide

H1071-25MG H1071-100MG
TCIAH1071-25MGEA 795 NOK
TCIAH1071-25MG TCIAH1071-100MG
Biotin-LC-hydrazide ≥97.0% (by HPLC)
Biotin-LC-hydrazide
Formel: C₁₆H₂₉N₅O₃S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00077659
CAS-nummer: 109276-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cb1f29269397464e1f852b58f72d92e81ef358b&vid=4cb1f29269397464e1f852b58f72d92e81ef358b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 18.00 to 19.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR