trans-2-Hexenyl valerate ≥94.0% (by GC)

Leverandør: TCI

Synonymer: Valeric Acid trans-2-Hexenyl Ester

H1444-25ML
TCIAH1444-25MLEA 3975 NOK
TCIAH1444-25ML
trans-2-Hexenyl valerate ≥94.0% (by GC)
trans-2-Hexenyl valerate
Formel: C₁₁H₂₀O₂
MW: 184,28 g/mol
MDL nummer: MFCD00036547
CAS-nummer: 56922-74-8
EINECS: 260-439-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c1bf45833aa859183d73d9d3dd6b5420aa68469&vid=6c1bf45833aa859183d73d9d3dd6b5420aa68469&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR