2H-Perfluoro-5-methyl-3,6-dioxanonane ≥90.0% (by GC)

Leverandør: TCI

Synonymer: 1,1,1,2,2,3,3-Heptafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy]propane

H1624-25G
TCIAH1624-25GEA 850 NOK
TCIAH1624-25G
2H-Perfluoro-5-methyl-3,6-dioxanonane ≥90.0% (by GC)
2H-Perfluoro-5-methyl-3,6-dioxanonane
Formel: C₈HF₁₇O₂
MW: 452,07 g/mol
Kokepunkt: 105°C
MDL nummer: MFCD00054714
CAS-nummer: 3330-14-1

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR