Inosine-5'-monophosphate barium salt hydrate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Barium Inosinate, 5'-IMP Ba, Inosic Acid Barium Salt, 5'-Inosinic Acid Barium Salt

I0039-100MG
TCIAI0039-100MGEA 745 NOK
TCIAI0039-100MG
Inosine-5'-monophosphate barium salt hydrate ≥95.0% (by titrimetric analysis)
Inosine-5'-monophosphate barium salt hydrate
Formel: C₁₀H₁₃N₄O₈P
MW: 483.52 g/mol
MDL nummer: MFCD00067321
CAS-nummer: 3249-92-1
FN: 1564
ADR: 6.1 III
Merck Indeks: 14,04977
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30c65d7600988329e11951fdf2c283cc01fa2d9f&vid=30c65d7600988329e11951fdf2c283cc01fa2d9f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 95.0 % (calcd.on anh.substance)
abs-ratio (a/b) 1.55 to 1.75 (E250/E260)
abs-ratio (2a/2b) 0.15 to 0.19 (E280/E260)
Water max. 22.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR