Isobutansyre ≥99.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-Methylpropionic acid, 2-Methylpropanoic acid

I0103-25ML I0103-500ML
TCIAI0103-25MLEA 220 NOK
TCIAI0103-25ML TCIAI0103-500ML
Isobutansyre ≥99.0% (by GC)
Isobutansyre
Formel: C₄H₈O₂
Kokepunkt: 155°C
Smeltepunkt: –47°C
Tetthet: 0,949 g/cm³ (20 °C)
Flash punkt: 56°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002658
CAS-nummer: 79-31-2
EINECS: 201-195-7
FN: 2529
ADR: 3,III
Merck Indeks: 14,05155
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c8c69aac5cb01cf5f3b8e1a74a3a5cef2eec862&vid=4c8c69aac5cb01cf5f3b8e1a74a3a5cef2eec862&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR