Isovaleric anhydride ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Isopentanoic anhydride

I0199-25ML I0199-500ML
TCIAI0199-25MLEA 380 NOK
TCIAI0199-25ML TCIAI0199-500ML
Isovaleric anhydride ≥99.0% (by titrimetric analysis)
Isovaleric anhydride
Formel: C₁₀H₁₈O₃
MW: 186.25 g/mol
Kokepunkt: 215°C
Smeltepunkt: -65
Flash punkt: 91°C
MDL nummer: MFCD00015051
CAS-nummer: 1468-39-9
EINECS: 215-994-3
FN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e17a340d45f79e289dbfd9bdffa425d1dd908e6&vid=8e17a340d45f79e289dbfd9bdffa425d1dd908e6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %
Purity (Morpholine Method) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR