Indophenol blue

Leverandør: TCI
Warning

Synonymer: N-(4-Dimethylaminophenyl)-1,4-naphthoquinoneimine, alpha-Naphthol Blue

I0208-1G
TCIAI0208-1GEA 1050 NOK
TCIAI0208-1G
Indophenol blue
Indophenol blue
Formel: C₁₈H₁₆N₂O
MW: 276,34 g/mol
MDL nummer: MFCD00041739
CAS-nummer: 132-31-0
EINECS: 205-056-1

Bestill nå

Spesifikasjon Testresultater

Appearance Gray to Dark purple to Black powder to crystal
Lambda max. 585.0 to 595.0 nm (EtOH)
Molar extinction coefficient min. 7500 (EtOH, 585.0 to 595.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR