Indigo carmine ≥95.0% (by UV/Vis)

Leverandør: TCI
Warning

Synonymer: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

I0214-25G
TCIAI0214-25GEA 365 NOK
TCIAI0214-25G
Indigo carmine ≥95.0% (by UV/Vis)
Indigo carmine
Formel: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005723
CAS-nummer: 860-22-0
EINECS: 212-728-8
Merck Indeks: 14,04944
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2dd9f3fc8c88e146fb4572bed382dcfbdd0d464&vid=e2dd9f3fc8c88e146fb4572bed382dcfbdd0d464&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (UV-vis method) min. 95.0 % (after drying)
Drying loss max. 8.0 %
Transition interval (pH) (blue)11.6 to 14.0 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR