Isopropanolamine phosphate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

I0221-25G
TCIAI0221-25GEA 1650 NOK
TCIAI0221-25G
Isopropanolamine phosphate ≥98.0% (by titrimetric analysis)
Isopropanolamine phosphate
Formel: C₃H₁₂NO₅P
MDL nummer: MFCD00070562
CAS-nummer: 67952-32-3
EINECS: 267-885-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR