Diphenyl isophthalate ≥99.0% (by GC)

Leverandør: TCI
Warning

I0416-10G I0416-25G
TCIAI0416-10GEA 555 NOK
TCIAI0416-10G TCIAI0416-25G
Diphenyl isophthalate ≥99.0% (by GC)
Diphenyl isophthalate
Formel: C₂₀H₁₄O₄
MDL nummer: MFCD00046051
CAS-nummer: 744-45-6
EINECS: 212-014-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 136.0 to 139.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR