(S,S,S)-Triglycidyl isocyanurate ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Danger

I0581-1G
TCIAI0581-1GEA 1225 NOK
TCIAI0581-1G
(S,S,S)-Triglycidyl isocyanurate ≥98.0% (by total nitrogen basis)
(S,S,S)-Triglycidyl isocyanurate
Formel: C₁₂H₁₅N₃O₆
MDL nummer: MFCD03844799
CAS-nummer: 240408-81-5
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=053fc6af72392a0fda310ae8785f8dc6e3fe2f5e&vid=053fc6af72392a0fda310ae8785f8dc6e3fe2f5e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Melting point 105.0 to 108.0 deg-C
Specific rotation [a]20/D -20.0 to -24.0 deg (C=0.4, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR