1L-epi-2-Inosose ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 2L-2,3,4,6/5-Pentahydroxycyclohexanone

I0634-200MG
TCIAI0634-200MGEA 810 NOK
TCIAI0634-200MG
1L-epi-2-Inosose ≥98.0% (by HPLC)
1L-epi-2-Inosose
Formel: C₆H₁₀O₆
MW: 178.14 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00038395
CAS-nummer: 33471-33-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -3.0 to -5.5 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR