Isorhapontigenin ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (E)-5-(4-hydroxy-3-methoxystyryl)benzene-1,3-diol, 3,4',5-Trihydroxy-3'-methoxy-trans-stilbene

I0804-100MG
TCIAI0804-100MGEA 1650 NOK
TCIAI0804-100MG
Isorhapontigenin ≥96.0% (by GC)
Isorhapontigenin
Formel: C₁₅H₁₄O₄
MW: 258,27 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD12407151
CAS-nummer: 32507-66-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 180.0 to 184.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR