4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

I0873-5G
TCIAI0873-5GEA 3975 NOK
TCIAI0873-5G
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₁H₂₀O₂
CAS-nummer: 38792-88-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e478ca3844e7db2b77d0697f5531eb4d40e562b4&vid=e478ca3844e7db2b77d0697f5531eb4d40e562b4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR