5-Iodo-2'-deoxycytidine 98,0 HPLC, T

Leverandør: TCI

Synonymer: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one

I0882-1G
TCIAI0882-1GEA 1625 NOK
TCIAI0882-1G
5-Iodo-2'-deoxycytidine 98,0 HPLC, T
5-Iodo-2'-deoxycytidine
Formel: C₉H₁₂IN₃O₄
MW: 353.12 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00038063
CAS-nummer: 611-53-0
EINECS: 210-269-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cbb0e4e91850d3b02cb3035fa6583b1fbb81a02&vid=6cbb0e4e91850d3b02cb3035fa6583b1fbb81a02&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 183.0 to 187.0 deg-C
Specific rotation [a]20/D +27.0 to +30.0 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR