2-Metylpropen ∼6% in toluene

Leverandør: TCI
Danger

Synonymer: 2-Methylpropene, Isobutene, Isobutylene

I1082-100ML I1082-500ML
TCIAI1082-100MLEA 340 NOK
TCIAI1082-100ML TCIAI1082-500ML
2-Metylpropen ∼6% in toluene
2-Metylpropen
Formel: C₄H₈
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008898
CAS-nummer: 115-11-7
EINECS: 204-066-3
FN: 1993
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 6.0 to 7.0 w/w%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR