2,6,6-Trimethyl-2-cyclohexene-1,4-dione ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Ketoisophorone, 4-Oxoisophorone

K0034-25G
TCIAK0034-25GEA 540 NOK
TCIAK0034-25G
2,6,6-Trimethyl-2-cyclohexene-1,4-dione ≥98.0% (by GC)
2,6,6-Trimethyl-2-cyclohexene-1,4-dione
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Kokepunkt: 214 °C (1013 hPa)
Smeltepunkt: 15 °C
Tetthet: 1,03 g/cm³ (20 °C)
MDL nummer: MFCD00043119
CAS-nummer: 1125-21-9
EINECS: 214-406-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55cbbcee4a8ada950d259259951e20a8870a5432&vid=55cbbcee4a8ada950d259259951e20a8870a5432&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR