Kojic acid sodium salt hydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one Sodium Salt

K0036-5G
TCIAK0036-5GEA 4475 NOK
TCIAK0036-5G
Kojic acid sodium salt hydrate ≥98.0% (by titrimetric analysis)
Kojic acid sodium salt hydrate
Formel: C₆H₆O₄
MW: 164.09 g/mol
MDL nummer: MFCD00191537
CAS-nummer: 70145-54-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 31.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR