Sodium-2-oxobutyrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 2-Oxobutyric Acid Sodium Salt, Sodium 2-Ketobutyrate, 2-Ketobutyric Acid Sodium Salt

K0040-5G
TCIAK0040-5GEA 1475 NOK
TCIAK0040-5G
Sodium-2-oxobutyrate ≥98.0% (by titrimetric analysis)
Sodium-2-oxobutyrate
Formel: C₄H₆O₃
MW: 124,07 g/mol
MDL nummer: MFCD00064194
CAS-nummer: 2013-26-5
EINECS: 217-937-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ef6ff94b996f51a76c60e5fe713c65fbf8cb250&vid=1ef6ff94b996f51a76c60e5fe713c65fbf8cb250&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water very faint turbidity
Water max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR