Calcium lactobionate hydrate ≥97.0% (by titrimetric analysis)

Leverandør: TCI

L0006-25G
TCIAL0006-25GEA 835 NOK
TCIAL0006-25G
Calcium lactobionate hydrate ≥97.0% (by titrimetric analysis)
Calcium lactobionate hydrate
Formel: C₁₂H₂₂O₁₂
MDL nummer: MFCD00135895
CAS-nummer: 5001-51-4
EINECS: 225-668-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +22.0 to +25.0 deg (C=6, H2O)
Water 2.0 to 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR