Lys Grønn SF Gulaktig

Leverandør: TCI

Synonymer: Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Light Green 2G, S or 2GN, CI 42095, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, CN DcL-30

L0054-25G
TCIAL0054-25GEA 1425 NOK
TCIAL0054-25G
Lys Grønn SF Gulaktig
Lys Grønn SF Gulaktig
Formel: C₃₇H₃₄N₂Na₂O₉S₃
Smeltepunkt: 288…289 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012121
CAS-nummer: 5141-20-8
EINECS: 225-906-5
Merck Indeks: 14,05485
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c403b28b5c16c58cf1b2de27c49b38e65cb5d630&vid=c403b28b5c16c58cf1b2de27c49b38e65cb5d630&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Lambda max. 631.0 to 635.0 nm (H2O)
Absorbance (E1%1cm) min. 700 (H2O, 631.0 to 635.0 nm)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR