Lithol rubin BCA 85,0 UV/Vis

Leverandør: TCI
Warning

Synonymer: Brilliant Carmine 6B, Pigment Red 57

L0075-25G
TCIAL0075-25GEA 475 NOK
TCIAL0075-25G
Lithol rubin BCA 85,0 UV/Vis
Lithol rubin BCA
Formel: C₁₈H₁₄N₂O₆S
MW: 424.44 g/mol
MDL nummer: MFCD00059529
CAS-nummer: 5281-04-9
EINECS: 226-109-5

Bestill nå

Spesifikasjon Testresultater

Appearance Red to Dark red to Brown powder to crystal
Purity (UV-vis method) min. 85.0 %
Lambda max. 518.0 to 522.0 nm (HCl (EtOH))


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR