L(+)-Lysine dihydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: H-Lys-OH·2HCl, H-Lys-OH.2HCl, (S)-(+)-Lysine dihydrochloride, (S)-(+)-2,6-Diaminohexanoic acid dihydrochloride, (S)-(+)-2,6-Diaminocaproic acid dihydrochloride

L0131-25G
TCIAL0131-25GEA 230 NOK
TCIAL0131-25G
L(+)-Lysine dihydrochloride ≥98.0% (by titrimetric analysis)
L(+)-Lysine dihydrochloride
Formel: C₆H₁₆Cl₂N₂O₂
MW: 219.11 g/mol
Smeltepunkt: 200.2
MDL nummer: MFCD00066389
CAS-nummer: 657-26-1
EINECS: 211-518-3
Merck Indeks: 14,05636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a393a7557ff98f81804e76c6d4b4ff598693bf00&vid=a393a7557ff98f81804e76c6d4b4ff598693bf00&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +13.0 to +16.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR