Lys Grønn SF Gulaktig indicator

Leverandør: TCI

Synonymer: Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Light Green 2G, S or 2GN, CI 42095, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, CN DcL-30

L0141-1G
TCIAL0141-1GEA 220 NOK
TCIAL0141-1G
Lys Grønn SF Gulaktig indicator
Lys Grønn SF Gulaktig

Formerly Food Green No.2, indicator for assay of Food Yellow No.4 (Tartrazine)

Formel: C₃₇H₃₄N₂Na₂O₉S₃
MDL nummer: MFCD00012121
CAS-nummer: 5141-20-8
EINECS: 225-906-5
Merck Indeks: 14,05485

Bestill nå

Spesifikasjon Testresultater

Appearance Red to Dark red to Dark purple powder to crystal
Lambda max. 630.0 to 634.0 nm (H2O)
Absorbance (E1%1cm) min. 700 (H2O, 630.0 to 634.0 nm)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR