L(+)-Lithium lactate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Lithium (S)-2-hydroxypropanoate, (S)-(+)-Lactic acid lithium salt, (S)-2-Hydroxypropionic acid Lithium salt, Lithium (S)-2-Hydroxypropionate, L(+)-Lactic acid lithium salt, (S)-(+)-2-Hydroxypropanoic acid lithium salt, (S)-(+)-2-Hydroxypropionic acid lithium salt, Lithium L-Lactate, L-Lactic acid Lithium salt

L0144-25G
TCIAL0144-25GEA 770 NOK
TCIAL0144-25G
L(+)-Lithium lactate ≥98.0% (by titrimetric analysis)
L(+)-Lithium lactate
Formel: C₃H₅LiO₃
MW: 96.01 g/mol
Kokepunkt: 200
Smeltepunkt: 75
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00065512
CAS-nummer: 27848-80-2
EINECS: 248-692-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33d2bf5a0f8f7a0ef3ab6780673ca4c113699fa2&vid=33d2bf5a0f8f7a0ef3ab6780673ca4c113699fa2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -12.0 to -15.0 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR