Dimethyl-DL-malate 97,0 GC_ASSAY_METHOD

Leverandør: TCI

Synonymer: DL-Apple Acid Dimethyl Ester, DL-Malic Acid Dimethyl Ester

M0025-10G
TCIAM0025-10GEA 825 NOK
TCIAM0025-10G
Dimethyl-DL-malate 97,0 GC_ASSAY_METHOD
Dimethyl-DL-malate
Formel: C₆H₁₀O₅
MW: 162.14 g/mol
Kokepunkt: 205°C
MDL nummer: MFCD00025868
CAS-nummer: 1587-15-1
EINECS: 216-448-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR