2-Merkaptoetanol ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

M0058-25G M0058-500G
TCIAM0058-25GEA 220 NOK
TCIAM0058-25G TCIAM0058-500G
2-Merkaptoetanol ≥98.0% (by GC)
2-Merkaptoetanol
Formel: HSCH₂CH₂OH
MW: 78.14 g/mol
Kokepunkt: 158°C
Smeltepunkt: –100°C
Flash punkt: 74°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004890
CAS-nummer: 60-24-2
EINECS: 200-464-6
FN: 2966
ADR: 6.1 II
Merck Indeks: 14,05869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b2de4a27b611bfba008542b0f8d01b41c1f5970&vid=4b2de4a27b611bfba008542b0f8d01b41c1f5970&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR