DL-Methionine sulphoxide ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 2-Amino-4-(methylsulphinyl)butanoic acid

M0101-5G
TCIAM0101-5GEA 1450 NOK
TCIAM0101-5G
DL-Methionine sulphoxide ≥98.0% (by titrimetric analysis)
DL-Methionine sulphoxide
Formel: C₅H₁₁NO₃S
MW: 165,21 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00002620
CAS-nummer: 62697-73-8
EINECS: 263-700-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5ad406149beae2d95212af659a00035c17dcffa&vid=c5ad406149beae2d95212af659a00035c17dcffa&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR