6-Methoxy-m-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

M0115-25G M0115-500G
TCIAM0115-25GEA 245 NOK
TCIAM0115-25G TCIAM0115-500G
6-Methoxy-m-toluidine ≥98.0% (by GC, titration analysis)
6-Methoxy-m-toluidine
Formel: C₈H₁₁NO
Kokepunkt: 235°C
Tetthet: 1 g/cm³ (20 °C)
MDL nummer: MFCD00007815
CAS-nummer: 120-71-8
EINECS: 204-419-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48205ce1615e67a46a0378902fd8ede9e9da7ff9&vid=48205ce1615e67a46a0378902fd8ede9e9da7ff9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 51.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR