Methylaminoacetonitrile sulphate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Warning

M0141-25G
TCIAM0141-25GEA 1600 NOK
TCIAM0141-25G
Methylaminoacetonitrile sulphate ≥95.0% (by titrimetric analysis)
Methylaminoacetonitrile sulphate
Formel: C₃H₆N₂
MW: 238,26 g/mol
MDL nummer: MFCD00035599
CAS-nummer: 33986-15-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 159.0 to 163.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR