3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: MCI-186, 1-Phenyl-3-methyl-5-pyrazolone, 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one, Edaravone

M0687-25G M0687-500G
TCIAM0687-25GEA 435 NOK
TCIAM0687-25G TCIAM0687-500G
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98.0% (by HPLC, titration analysis)
3-Methyl-1-phenyl-2-pyrazolin-5-one
Formel: C₁₀H₁₀N₂O
MDL nummer: MFCD00003138
CAS-nummer: 89-25-8
EINECS: 201-891-0
Merck Indeks: 14,06713
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9295eb4bfa90f659439971706118b3c946b7d9b&vid=d9295eb4bfa90f659439971706118b3c946b7d9b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 131.0 deg-C
Solubility in N,N-DMF almost transparency

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR