6-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

M0772-25ML M0772-500ML
TCIAM0772-25MLEA 220 NOK
TCIAM0772-25ML TCIAM0772-500ML
6-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)
6-Ethyl-o-toluidine
Formel: C₉H₁₃N
Kokepunkt: 231 °C (1013 hPa)
Smeltepunkt: –33 °C
Tetthet: 0,969 g/cm³ (20 °C)
Flash punkt: 111°C
MDL nummer: MFCD00007752
CAS-nummer: 24549-06-2
EINECS: 246-309-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52dcee054d2eeaa672f848513160ae6e16bd8b95&vid=52dcee054d2eeaa672f848513160ae6e16bd8b95&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR