4-(Dimethylamino)-2'-methylazobenzene

Leverandør: TCI
Warning

Synonymer: 2'-Methyl-4-(dimethylamino)azobenzene, 2-Methyl-4-dimethylaminoazobenzene

M0789-1G
TCIAM0789-1GEA 1325 NOK
TCIAM0789-1G
4-(Dimethylamino)-2'-methylazobenzene
4-(Dimethylamino)-2'-methylazobenzene
Formel: C₁₅H₁₇N₃
MW: 239,32 g/mol
Smeltepunkt: 74°C
MDL nummer: MFCD00025648
CAS-nummer: 3731-39-3
EINECS: 223-088-4

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Amber to Dark red powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR