1H-1,2,4-Triazole-3-thiol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

M0814-25G M0814-500G M0814-100G
TCIAM0814-25GEA 460 NOK
TCIAM0814-25G TCIAM0814-500G TCIAM0814-100G
1H-1,2,4-Triazole-3-thiol ≥98.0% (by GC, titration analysis)
1H-1,2,4-Triazole-3-thiol
Formel: C₂H₃N₃S
Smeltepunkt: 217…220 °C
MDL nummer: MFCD00005229
CAS-nummer: 3179-31-5
EINECS: 221-656-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cefe2c3664fa86008cfb4123040760bbe65f072&vid=0cefe2c3664fa86008cfb4123040760bbe65f072&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 218.0 to 223.0 deg-C
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR