L-Methionine methyl ester hydrochloride ≥96.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: H-Met-OMe.HCl, H-Met-OMe·HCl

M0853-5G
TCIAM0853-5GEA 350 NOK
TCIAM0853-5G
L-Methionine methyl ester hydrochloride ≥96.0% (by titrimetric analysis)
L-Methionine methyl ester hydrochloride
Formel: C₆H₁₃NO₂S·HCl
MW: 199,7 g/mol
MDL nummer: MFCD00012491
CAS-nummer: 2491-18-1
EINECS: 219-651-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea1e05e15cd531703124fe2f837cc2a4bd2e5694&vid=ea1e05e15cd531703124fe2f837cc2a4bd2e5694&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 96.0 %
Specific rotation [a]20/D +25.0 to +27.0 deg (C=1, H2O)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR