Methylcyclopentadiene dimer (so called) stabilised

Leverandør: TCI
Warning


Stabilisering: stabilisert med 4-tert-Butylpyrokatekol

M0920-25ML
TCIAM0920-25MLEA 220 NOK
TCIAM0920-25ML
Methylcyclopentadiene dimer (so called) stabilised
Methylcyclopentadiene dimer
Formel: C₁₂H₁₆
MW: 160,26 g/mol
Flash punkt: 53°C
MDL nummer: MFCD00082297
CAS-nummer: 26472-00-4
EINECS: 247-724-5
FN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=601be4e54f206e7ee8802309a01d27085885d02a&vid=129aafdf63318305f6a139a96022d46e78b2799c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR