N-Methyl-4-nitro-o-toluidine ≥97.0% (by GC)

Leverandør: TCI
Warning

M0953-5G
TCIAM0953-5GEA 5150 NOK
TCIAM0953-5G
N-Methyl-4-nitro-o-toluidine ≥97.0% (by GC)
N-Methyl-4-nitro-o-toluidine
Formel: C₈H₁₀N₂O₂
MDL nummer: MFCD00059083
CAS-nummer: 10439-77-7

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 137.0 to 141.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR