2-Mercapto-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

M0976-25G
TCIAM0976-25GEA 420 NOK
TCIAM0976-25G
2-Mercapto-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)
2-Mercapto-5-methyl-1,3,4-thiadiazole
Formel: C₃H₄N₂S₂
MDL nummer: MFCD00038349
CAS-nummer: 29490-19-5
EINECS: 249-667-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d2474d5a41c1f5175d58da4bda8efc0ee1ca39e&vid=4d2474d5a41c1f5175d58da4bda8efc0ee1ca39e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 184.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR