Methyl-β-D-arabinopyranoside ≥99.0% (by GC)

Leverandør: TCI

M1019-100MG
TCIAM1019-100MGEA 555 NOK
TCIAM1019-100MG
Methyl-β-D-arabinopyranoside ≥99.0% (by GC)
Methyl-beta-D-arabinopyranoside
Formel: C₆H₁₂O₅
MW: 164,16 g/mol
MDL nummer: MFCD00063261
CAS-nummer: 5328-63-2
EINECS: 226-215-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 171.0 to 175.0 deg-C
Specific rotation [a]20/D -235.0 to -245.0 deg (C=7, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR