2-[2-(5-Methylfuran-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine ≥85.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 2,4-Bis(trichloromethyl)-6-[2-(5-methylfuran-2-yl)vinyl]-1,3,5-triazine

M1209-5G
TCIAM1209-5GEA 1125 NOK
TCIAM1209-5G
2-[2-(5-Methylfuran-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine ≥85.0% (by HPLC, titration analysis)
2-[2-(5-Methylfuran-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Formel: C₁₂H₇Cl₆N₃O
MW: 421,91 g/mol
MDL nummer: MFCD00671558
CAS-nummer: 156360-76-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 85.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR