2-Mercapto-5-methoxybenzimidazole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

M1256-25G
TCIAM1256-25GEA 715 NOK
TCIAM1256-25G
2-Mercapto-5-methoxybenzimidazole ≥98.0% (by HPLC, titration analysis)
2-Mercapto-5-methoxybenzimidazole
Formel: C₈H₈N₂OS
Kokepunkt: 400 °C (1013 hPa)
Smeltepunkt: 261…263 °C
MDL nummer: MFCD00134581
CAS-nummer: 37052-78-1
EINECS: 253-326-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b586838ed3a152a30888c86cbd23864d902247d5&vid=b586838ed3a152a30888c86cbd23864d902247d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR