3-Methylpent-1-en-4-yn-3-ol ≥98.0% (by GC)

Leverandør: TCI
Warning

M1312-5ML
TCIAM1312-5MLEA 875 NOK
TCIAM1312-5ML
3-Methylpent-1-en-4-yn-3-ol ≥98.0% (by GC)
3-Methylpent-1-en-4-yn-3-ol
Formel: C₆H₈O
MW: 96,13 g/mol
Kokepunkt: 121°C
Flash punkt: 32°C
MDL nummer: MFCD01709006
CAS-nummer: 3230-69-1
EINECS: 221-770-6
FN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbb7dfb647912307c352002513e71ff30de26a0a&vid=fbb7dfb647912307c352002513e71ff30de26a0a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR