(+)-Dimethyl-D-malate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: D-(+)-Malic acid dimethyl ester, Dimethyl D-(+)-Malate, D-(+)-Apple acid dimethyl ester, (R)-Dimethyl 2-hydroxysuccinate

M1347-25G M1347-5G
TCIAM1347-25GEA 4100 NOK
TCIAM1347-25G TCIAM1347-5G
(+)-Dimethyl-D-malate ≥98.0% (by GC)
(+)-Dimethyl-D-malate
Formel: C₆H₁₀O₅
MW: 162.1406 g/mol
MDL nummer: MFCD00066216
CAS-nummer: 70681-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf7c34028c7bd74a1ffa8424cad28c7894089e05&vid=bf7c34028c7bd74a1ffa8424cad28c7894089e05&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Specific rotation [a]20/D +5.0 to +8.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR