1-Methoxy-5-methylphenazinium methyl sulphate ≥95.0% (by HPLC) for biochemical research

Leverandør: TCI
Warning

M1442-200MG
TCIAM1442-200MGEA 1350 NOK
TCIAM1442-200MG
1-Methoxy-5-methylphenazinium methyl sulphate ≥95.0% (by HPLC) for biochemical research
1-Methoxy-5-methylphenazinium methyl sulphate
Formel: C₁₅H₁₆N₂O₅S
Smeltepunkt: 172°C
MDL nummer: MFCD00040648
CAS-nummer: 65162-13-2
EINECS: 265-579-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c867fb52577541e2524d7db0671bfcacff079a7d&vid=38a94511bbdd91801019dd2d1027bb266fde37ac&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 95.0 area%
Enzyme detection test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR