Methylenebis[dimethyl(2-pyridyl)silane] ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,2'-[Methylenebis(dimethylsilylene)]bispyridine, Bis[dimethyl(2-pyridyl)silyl]methane

M1460-100MG
TCIAM1460-100MGEA 6975 NOK
TCIAM1460-100MG
Methylenebis[dimethyl(2-pyridyl)silane] ≥95.0% (by GC)
Methylenebis[dimethyl(2-pyridyl)silane]
Formel: C₁₅H₂₂N₂Si₂
MW: 286,53 g/mol
MDL nummer: MFCD06797120
CAS-nummer: 243468-48-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR