2-(N-Methyl-2-pyridylamino)ethanol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-(Methyl(pyridin-2-yl)amino)ethanol, N-Methyl-N-(2-pyridyl)ethanolamine, 2-[N-(2-Hydroxyethyl)-N-methylamino]pyridine

M1468-5G
TCIAM1468-5GEA 325 NOK
TCIAM1468-5G
2-(N-Methyl-2-pyridylamino)ethanol ≥98.0% (by GC, titration analysis)
2-(N-Methyl-2-pyridylamino)ethanol
Formel: C₈H₁₂N₂O
MW: 152,20 g/mol
MDL nummer: MFCD02667633
CAS-nummer: 122321-04-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR