4-Methoxyphenyl-2,3,4,6-tetra-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)

Leverandør: TCI

M1588-5G
TCIAM1588-5GEA 2250 NOK
TCIAM1588-5G
4-Methoxyphenyl-2,3,4,6-tetra-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,3,4,6-tetra-O-benzyl-beta-D-galactopyranoside
Formel: C₄₁H₄₂O₇
MW: 646,78 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD06797131
CAS-nummer: 143536-99-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 108.0 to 112.0 deg-C
Specific rotation [a]20/D -16.0 to -19.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR