4-Methoxyphenyl-2,4,6-tri-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)

Leverandør: TCI

M1592-1G
TCIAM1592-1GEA 2175 NOK
TCIAM1592-1G
4-Methoxyphenyl-2,4,6-tri-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,4,6-tri-O-benzyl-beta-D-galactopyranoside
Formel: C₃₄H₃₆O₇
MW: 556,66 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD06797134
CAS-nummer: 247027-79-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 92.0 to 96.0 deg-C
Specific rotation [a]20/D -13.0 to -16.0 deg (C=1, CHCl3)
Solubility in Chloroform very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR