4-Methylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

M1621-25G M1621-5G
TCIAM1621-25GEA 2600 NOK
TCIAM1621-25G TCIAM1621-5G
4-Methylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-Methylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₈H₁₄O₂
Kokepunkt: 140°C
MDL nummer: MFCD00074909
CAS-nummer: 4331-54-8
EINECS: 224-369-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR